Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethanethiol, 95%

Catalog No. AC434710050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434710050 5 g Glass bottle
AC434710250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434710050 Supplier Thermo Scientific Chemicals Supplier No. 434710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 60-23-1 | C2H7NS | 77.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-23-1
Molecular Formula C2H7NS
Molecular Weight (g/mol) 77.15
MDL Number MFCD00008196
InChI Key UFULAYFCSOUIOV-UHFFFAOYSA-N
Synonym cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten
PubChem CID 6058
ChEBI CHEBI:17141
IUPAC Name 2-aminoethanethiol
SMILES NCCS

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 95.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Linear Formula NH2CH2CH2SH
Quantity 5 g
Merck Index 15,2776
Solubility Information Solubility in water: freely soluble.
Formula Weight 77.15
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Aminoethanethiol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:


WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-463-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.