Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethanethiol 95.0+%, TCI America™
SDP

Catalog No. A064825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A064825G 25 g
A0648500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A064825G Supplier TCI America Supplier No. A064825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-23-1
Molecular Formula C2H7NS
Molecular Weight (g/mol) 77.15
MDL Number MFCD00008196
InChI Key UFULAYFCSOUIOV-UHFFFAOYSA-N
Synonym cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten
PubChem CID 6058
ChEBI CHEBI:17141
IUPAC Name 2-aminoethane-1-thiol
SMILES NCCS

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
Formula Weight 77.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanethiol
EINECSNumber (9)-1422
RTECSNumber KJ0175000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.