Learn More
TARGETMOL CHEMICALS INC 2-AMINOETHYL DIPHENYLBOR 50MG
SDP
Catalog No. 502576733
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 2-Aminoethyl diphenylborinate (2-APB) is a chemical that acts to inhibit both IP3 receptors and TRP channels (although it activates TRPV1 TRPV2 & TRPV3 at higher concentrations). purity: 99%
Catalog No. 50-257-6733 Supplier TARGETMOL CHEMICALS INC Supplier No. T469350MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 2-Aminoethyl diphenylborinate (2-APB) is a chemical that acts to inhibit both IP3 receptors and TRP channels (although it activates TRPV1 TRPV2 & TRPV3 at higher concentrations). purity: 99%