Learn More
Medchemexpress LLC 2-Aminoethyl diphenylborinate (2-APB) | 524-95-8 | 98.9% | 225.09 | 10 MG
SDP
Catalog No. 5000453343
Encompass_Preferred
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R). It also inhibits the store-operated Ca2+ (SOC) channel and activates some TRP channels (V1, V2 and V3). In addition, it has inhibitory effects on vasospasm. At high concentrations, it demonstrates specific anti-inflammatory and antioxidant effects in neural tissue.
  • Cell-permeable inhibitor of inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channel
  • Activates some TRP channels (V1, V2 and V3)
  • Exhibits inhibitory effects on vasospasm
  • Displays specific anti-inflammatory and antioxidant effects in neural tissue at high concentrations
Catalog No. 50-004-53343 Supplier Medchemexpress LLC Supplier No. HYW00972410MG
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R). It also inhibits the store-operated Ca2+ (SOC) channel and activates some TRP channels (V1, V2 and V3). In addition, it has inhibitory effects on vasospasm. At high concentrations, it demonstrates specific anti-inflammatory and antioxidant effects in neural tissue.
  • Cell-permeable inhibitor of inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channel
  • Activates some TRP channels (V1, V2 and V3)
  • Exhibits inhibitory effects on vasospasm
  • Displays specific anti-inflammatory and antioxidant effects in neural tissue at high concentrations