Learn More
Medchemexpress LLC 2-Aminoethyl diphenylborinate | 524-95-8 | 98.9% | 225.09 | 100 MG
SDP
Catalog No. 5000451592
Encompass_Preferred
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R). It also inhibits the store-operated Ca2+ (SOC) channel and activates some TRP channels (V1, V2, and V3). Additionally, it has inhibitory effects on vasospasm and exhibits specific anti-inflammatory and antioxidant effects in neural tissue at high concentrations. This product is for research use only.
  • Cell-permeable inhibitor of Inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channel
  • Activates TRP channels (V1, V2, and V3)
  • Inhibitory effects on vasospasm
  • Anti-inflammatory and antioxidant effects in neural tissue
Catalog No. 50-004-51592 Supplier Medchemexpress LLC Supplier No. HYW009724100MG
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R). It also inhibits the store-operated Ca2+ (SOC) channel and activates some TRP channels (V1, V2, and V3). Additionally, it has inhibitory effects on vasospasm and exhibits specific anti-inflammatory and antioxidant effects in neural tissue at high concentrations. This product is for research use only.
  • Cell-permeable inhibitor of Inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channel
  • Activates TRP channels (V1, V2, and V3)
  • Inhibitory effects on vasospasm
  • Anti-inflammatory and antioxidant effects in neural tissue