Learn More
Medchemexpress LLC 2-Aminoethyl diphenylborinate | 524-95-8 | 98.9% | 225.09 | 50 MG
SDP
Catalog No. 5000451593
Encompass_Preferred
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R) that also inhibits store-operated Ca2+ (SOC) channels and activates certain TRP channels. This compound has demonstrated inhibitory effects on vasospasm and, at high concentrations, exhibits specific anti-inflammatory and antioxidant effects in neural tissue. It is intended for research use only.
  • Cell-permeable inhibitor of Inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channels
  • Activates some TRP channels (V1, V2, and V3)
  • Exhibits inhibitory effects on vasospasm
  • Provides anti-inflammatory and antioxidant effects in neural tissue
  • Offers protective effects against ischemia-reperfusion induced damage
  • Reduces oxidative stress and apoptosis from testicular ischemia-reperfusion injury
Catalog No. 50-004-51593 Supplier Medchemexpress LLC Supplier No. HYW00972450MG
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R) that also inhibits store-operated Ca2+ (SOC) channels and activates certain TRP channels. This compound has demonstrated inhibitory effects on vasospasm and, at high concentrations, exhibits specific anti-inflammatory and antioxidant effects in neural tissue. It is intended for research use only.
  • Cell-permeable inhibitor of Inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channels
  • Activates some TRP channels (V1, V2, and V3)
  • Exhibits inhibitory effects on vasospasm
  • Provides anti-inflammatory and antioxidant effects in neural tissue
  • Offers protective effects against ischemia-reperfusion induced damage
  • Reduces oxidative stress and apoptosis from testicular ischemia-reperfusion injury