Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Aminoethyl Diphenylborinate

Catalog No. D975410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
25 g
5 Vial
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D975410G 1 g
D97541G 5 g
D97545G 25 g
D975425G 5 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. D975410G Supplier Sigma Aldrich Supplier No. D975410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of apoptotic inducersActivator of the Orai3 (ORAI calcium release-activated calcium modulator 3) proteinInhibits Ca2+ influx through transient receptor potential 2 channels in rat dorsal root ganglion neuronsMembrane-permeable SOCE (store operated calcuym entry) inhibitorMembrane-permeable modulator of intracellular IP3-induced calcium release.

Specifications

CAS 524-95-8
Molecular Formula C14H16BNO
Linear Formula (C6H5)2BOCH2CH2NH2
MDL Number MFCD00014823
Quantity 1 g
Synonym 2-APB; 2-Aminoethoxydiphenyl borate; 2-Aminoethyl diphenyl borate; Diphenylboric acid 2-aminoethyl ester; Diphenylborinic acid; NSC 17107
Molecular Weight (g/mol) 225.09 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.