Learn More
Medchemexpress LLC 2-Aminoethyl-mono-amide-DOTA-tris(tBu ester) | 173308-19-5 | MFCD30533300 | 98.0% | 614.82 g/mol | C30H58N6O7 | 1 G
SDP
Catalog No. 5000215828
Encompass_Preferred
2-Aminoethyl-mono-amide-DOTA-tris(tBu ester) is a protected DOTA-based metal chelator precursor used to prepare radiolabeled targeting agents for imaging and therapy. The tert-butyl ester groups protect DOTA carboxyls during synthesis and can be removed to expose carboxyls for conjugation to antibodies, peptides, or other targeting molecules. It is primarily intended for tumor pre-targeting and radionuclide research.
Catalog No. 50-002-15828 Supplier Medchemexpress LLC Supplier No. HY1001381G
2-Aminoethyl-mono-amide-DOTA-tris(tBu ester) is a protected DOTA-based metal chelator precursor used to prepare radiolabeled targeting agents for imaging and therapy. The tert-butyl ester groups protect DOTA carboxyls during synthesis and can be removed to expose carboxyls for conjugation to antibodies, peptides, or other targeting molecules. It is primarily intended for tumor pre-targeting and radionuclide research.