Learn More
Medchemexpress LLC 2-aminoethyl-mono-amide-DOTA-tris(tBu ester) | 173308-19-5 | MFCD30533300 | 98.0% | 614.82 g/mol | C30H58N6O7 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY100138250MG
Encompass_Preferred
2-Aminoethyl-mono-amide-DOTA-tris(tBu ester) is a protected DOTA-based chelator precursor supplied as a solid for research use in radiochemistry and targeted radiotherapy development. The tert-butyl ester groups protect carboxylates during synthesis and allow deprotection to yield a DOTA chelator that forms stable complexes with radiometals such as 68Ga and 177Lu for imaging and therapy applications.
  • Protected DOTA chelator precursor for radiometal labeling.
  • Compatible with conjugation to antibodies, peptides, and small molecules.
  • Enables stable complexation with common radiometals (for example, 68Ga and 177Lu).
  • High purity (98.0%) suitable for research applications.
  • Solid form for convenient handling and storage.
  • Useful in tumor pre-targeting and radiochemistry workflows.
Catalog No. 50-002-15221