Learn More
Medchemexpress LLC (2-Aminoethyl)phosphonic acid | 2041-14-7 | MFCD00008182 | 125.06 | 250 MG
SDP
Catalog No. 5000449797
Encompass_Preferred
(2-Aminoethyl)phosphonic acid is an endogenous metabolite and amino acid analog. It can be utilized for the synthesis of biological macromolecules and the preparation of hydrogels. In vivo, it has been observed to accumulate in various tissues such as the liver, kidney, and spleen after oral administration or intraperitoneal injection in rats.
  • Endogenous metabolite and amino acid analog
  • Utilized for the synthesis of biological macromolecules
  • Used for the preparation of hydrogels
  • Accumulates in various tissues such as the liver, kidney, and spleen after oral administration or intraperitoneal injection in rats.
Catalog No. 50-004-49797 Supplier Medchemexpress LLC Supplier No. 250MGHYW006371
(2-Aminoethyl)phosphonic acid is an endogenous metabolite and amino acid analog. It can be utilized for the synthesis of biological macromolecules and the preparation of hydrogels. In vivo, it has been observed to accumulate in various tissues such as the liver, kidney, and spleen after oral administration or intraperitoneal injection in rats.
  • Endogenous metabolite and amino acid analog
  • Utilized for the synthesis of biological macromolecules
  • Used for the preparation of hydrogels
  • Accumulates in various tissues such as the liver, kidney, and spleen after oral administration or intraperitoneal injection in rats.