Learn More
Selleck Chemical LLC 2-Aminoethylphosphonic acid S3331-5mg
SDP
Supplier:  Selleck Chemical LLC S33315MG
Encompass_Preferred
2-Aminoethylphosphonic acid (2-AEP (2-Aminoethyl)phosphonic acid) is a type of abundant and ubiquitous naturally occurring phosphonate used as sources of phosphorus by many prokaryotic lineages
Catalog No. 50-311-0462