Learn More
MOLTOX Molecular Toxicology Inc 2-Aminofluorene, 100 ug/vial, 5 vials/pack, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60104
Encompass
For use in bacterial reverse mutation assays with all Ames strains as well as WP2 tester strains. Dose 10 - 20 µg/plate. Reconstitute with organic solvents such as DMSO or sterile dH2O as applicable. Indirect acting mutagens require the use of S9 metabolic activation to cause mutagenicity. CONTROLCHEM™ chemicals are obtained from major suppliers of research chemicals and are employed without further characterization or purification. Packaged quantities are precise within 1%. Please Note: CONTROLCHEM™ chemicals are known mutagens/carcinogens/toxins and are sold only to those experienced in the safe handling and disposal of hazardous materials. Consult your Institutional Safety Officer before ordering.
Catalog No. 50-948-875