Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminofluorene | 153-78-6 | MFCD00001125 | 99.9% | 181.24 g·mol⁻¹ | C13H11N | 10 G
SDP

Catalog No. 5000302203
Encompass_Preferred

2-Aminofluorene is an aromatic amine used as a research reagent in studies of DNA adduct structure, DNA repair, carcinogenesis, and mutagenesis. It is supplied as a solid with high listed purity and defined storage conditions. The compound is a known genotoxin and is intended for research use only.

  • High purity: 99.9%.
  • Molecular formula: C13H11N; molecular weight: 181.24 g·mol⁻¹.
  • Suitable for research into DNA adduct formation and mutagenesis.
  • Available in solid form with multiple pack sizes, including a 10 g variant.
  • Off-white to light yellow solid for straightforward handling and characterization.
  • Storage stability: powder stored at -20°C (3 years) or 4°C (2 years).
  • Hazard: classified as a genotoxin; handle with appropriate safety precautions.

Catalog No. 50-003-02203 Supplier Medchemexpress LLC Supplier No. HYW01643310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminofluorene is an aromatic amine used as a research reagent in studies of DNA adduct structure, DNA repair, carcinogenesis, and mutagenesis. It is supplied as a solid with high listed purity and defined storage conditions. The compound is a known genotoxin and is intended for research use only.

  • High purity: 99.9%.
  • Molecular formula: C13H11N; molecular weight: 181.24 g·mol⁻¹.
  • Suitable for research into DNA adduct formation and mutagenesis.
  • Available in solid form with multiple pack sizes, including a 10 g variant.
  • Off-white to light yellow solid for straightforward handling and characterization.
  • Storage stability: powder stored at -20°C (3 years) or 4°C (2 years).
  • Hazard: classified as a genotoxin; handle with appropriate safety precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.