Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminofluorene | 153-78-6 | MFCD00001125 | 99.9% | 181.24 g/mol | C13H11N | 50 G
SDP

Catalog No. 5000299344
Encompass_Preferred

2-Aminofluorene is an aromatic amine used as a research-grade genotoxin for studies of DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. Supplied as a solid chemical standard, it is intended for laboratory research applications and is accompanied by product specifications and supporting documentation.

  • Research-grade genotoxin for DNA adduct, DNA repair, carcinogenesis, and mutagenesis studies.
  • High purity (manufacturer-stated 99.90%).
  • Molecular formula C13H11N and molecular weight 181.24 g/mol.
  • CAS number 153-78-6 for unambiguous identification.
  • Available in multiple pack sizes, including a 50 g option.
  • Provided with a downloadable product datasheet for technical reference.

Catalog No. 50-002-99344 Supplier Medchemexpress LLC Supplier No. HYW01643350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminofluorene is an aromatic amine used as a research-grade genotoxin for studies of DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. Supplied as a solid chemical standard, it is intended for laboratory research applications and is accompanied by product specifications and supporting documentation.

  • Research-grade genotoxin for DNA adduct, DNA repair, carcinogenesis, and mutagenesis studies.
  • High purity (manufacturer-stated 99.90%).
  • Molecular formula C13H11N and molecular weight 181.24 g/mol.
  • CAS number 153-78-6 for unambiguous identification.
  • Available in multiple pack sizes, including a 50 g option.
  • Provided with a downloadable product datasheet for technical reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.