Learn More
Medchemexpress LLC 2-aminofluorene-d11 | 347841-44-5 | 99.0% | 192.30 g·mol⁻¹ | C13D11N | 5 MG
SDP
Catalog No. 5000303320
Encompass_Preferred
2-Aminofluorene-d11 is the deuterium-labeled analogue of 2-aminofluorene supplied as a powder for use as an isotopic internal standard, tracer, or reference material in analytical and biochemical research. It is intended for applications that require mass-shifted analogues for mass spectrometry, metabolic studies, or DNA-adduct investigations.
  • Deuterium-labeled internal standard suitable for mass spectrometry.
  • High isotopic enrichment (98%) for clear mass separation.
  • High chemical purity (99% by HPLC) for analytical reliability.
  • Molecular weight supports precise mass calculations (192.30 g·mol⁻¹).
  • Supplied as a stable powder for ease of storage and handling.
Catalog No. 50-003-03320 Supplier Medchemexpress LLC Supplier No. HYW016433S15MG
2-Aminofluorene-d11 is the deuterium-labeled analogue of 2-aminofluorene supplied as a powder for use as an isotopic internal standard, tracer, or reference material in analytical and biochemical research. It is intended for applications that require mass-shifted analogues for mass spectrometry, metabolic studies, or DNA-adduct investigations.
  • Deuterium-labeled internal standard suitable for mass spectrometry.
  • High isotopic enrichment (98%) for clear mass separation.
  • High chemical purity (99% by HPLC) for analytical reliability.
  • Molecular weight supports precise mass calculations (192.30 g·mol⁻¹).
  • Supplied as a stable powder for ease of storage and handling.