Learn More
Medchemexpress LLC 2-aminofluorene-d11 | 347841-44-5 | MFCD01074176 | 99.0% | 192.3 g/mol | C13D11N | 1 MG
SDP
Catalog No. 5000303197
Encompass_Preferred
2-Aminofluorene-d11 is the deuterium-labeled isotopologue of 2-aminofluorene (CAS 347841-44-5), provided as a stable-isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, method development, and isotopic tracer studies; reported purity (HPLC) is 99% with isotopic enrichment around 98%.
  • Deuterium-labeled (d11) isotopologue for mass spectrometry.
  • High purity suitable for analytical standards and reference use.
  • High isotopic enrichment for reliable quantitation and tracing.
  • Supplied in small, weighed quantities for laboratory use.
  • Useful for method development, calibration, and isotope tracing studies.
Catalog No. 50-003-03197 Supplier Medchemexpress LLC Supplier No. HYW016433S11MG
2-Aminofluorene-d11 is the deuterium-labeled isotopologue of 2-aminofluorene (CAS 347841-44-5), provided as a stable-isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, method development, and isotopic tracer studies; reported purity (HPLC) is 99% with isotopic enrichment around 98%.
  • Deuterium-labeled (d11) isotopologue for mass spectrometry.
  • High purity suitable for analytical standards and reference use.
  • High isotopic enrichment for reliable quantitation and tracing.
  • Supplied in small, weighed quantities for laboratory use.
  • Useful for method development, calibration, and isotope tracing studies.