Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Aminoheptane | 123-82-0 | 99.8% | 500 MG
SDP

Catalog No. 5000407640
Encompass_Preferred

2-Aminoheptane, also known as Tuaminoheptane, is an isomeric heptylamine commonly used as a stimulant. It can also be utilized as a nitrogen source in minimal medium. This product is a colorless to light yellow liquid.

  • Isomeric heptylamine
  • Commonly used as a stimulant
  • Utilized as a nitrogen source in minimal medium
  • Appearance: colorless to light yellow liquid
  • Density: 0.766 g/cm³
  • Purity: 99.8%
  • Molecular Weight: 115.22
  • Molecular Formula: C7H17N
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-07640 Supplier Medchemexpress LLC Supplier No. HYB0952500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminoheptane, also known as Tuaminoheptane, is an isomeric heptylamine commonly used as a stimulant. It can also be utilized as a nitrogen source in minimal medium. This product is a colorless to light yellow liquid.

  • Isomeric heptylamine
  • Commonly used as a stimulant
  • Utilized as a nitrogen source in minimal medium
  • Appearance: colorless to light yellow liquid
  • Density: 0.766 g/cm³
  • Purity: 99.8%
  • Molecular Weight: 115.22
  • Molecular Formula: C7H17N
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.