Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Aminoheptane

Catalog No. A5620525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5620525G 25 g
A56205100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5620525G Supplier Sigma Aldrich Supplier No. A5620525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Aminoheptane is a secondary alkyl amine that can be used to synthesize 3-amino-2-heptanone hydrochloride.

Specifications

CAS 123-82-0
Density 0.766 g/mL (at 25°C (literature))
Boiling Point 142°C to 144°C (lit.)
Molecular Formula C7H17N
Refractive Index n20/D 1.418 (literature)
Linear Formula CH3(CH2)4 CH(NH2)CH3
MDL Number MFCD00008101
Quantity 25 g
Synonym 1-Methylhexylamine; 2-Heptylamine
Molecular Weight (g/mol) 115.22
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.