Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Aminoheptane sulfate, 99%, Thermo Scientific™

Catalog No. AAL0859622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0859622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0859622 Supplier Thermo Scientific Chemicals Supplier No. L0859622

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6411-75-2
Molecular Formula C14H36N2O4S
Molecular Weight (g/mol) 328.512
MDL Number MFCD00050844
InChI Key XKUUMWKWUZRRPD-UHFFFAOYSA-N
Synonym sulfuric acid; bis tuaminoheptane, dsstox_cid_25845, dsstox_rid_81167, dsstox_gsid_45845, -2-aminoheptane sulfate, tuaminoheptane sulfate usp, heptan-2-amine sulfate 2:1, ^+-2-aminoheptane sulfate
PubChem CID 44134533
IUPAC Name heptan-2-amine;sulfuric acid
SMILES CCCCCC(C)N.CCCCCC(C)N.OS(=O)(=O)O

Specifications

Melting Point 289°C to 291°C
Quantity 100 g
Beilstein 3703311
Merck Index 14,9802
Formula Weight 328.51
Percent Purity 99%
Chemical Name or Material (+/-)-2-Aminoheptane sulfate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 229-113-5
RTECSNumber MQ5600000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.