Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminoimidazole | 7720-39-0 | 98.1% | 83.09 g/mol | C3H5N3 | 250 MG
SDP

Catalog No. 5000209288
Encompass_Preferred

2-Aminoimidazole is a small-molecule antibiofilm agent used in research to inhibit bacterial biofilm formation and related resistance traits. It is provided in high purity for laboratory studies and is available in multiple pack sizes to support different experimental needs.

  • High purity (98.1%) for reliable experimental results.
  • Available in multiple pack sizes, including 250 MG.
  • Useful as an adjuvant to study antibiotic potentiation and biofilm disruption.
  • Chemical formula C3H5N3 and molecular weight 83.09 g/mol.
  • CAS number 7720-39-0 for unambiguous identification.

Catalog No. 50-002-09288 Supplier Medchemexpress LLC Supplier No. 250MGHYW062216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminoimidazole is a small-molecule antibiofilm agent used in research to inhibit bacterial biofilm formation and related resistance traits. It is provided in high purity for laboratory studies and is available in multiple pack sizes to support different experimental needs.

  • High purity (98.1%) for reliable experimental results.
  • Available in multiple pack sizes, including 250 MG.
  • Useful as an adjuvant to study antibiotic potentiation and biofilm disruption.
  • Chemical formula C3H5N3 and molecular weight 83.09 g/mol.
  • CAS number 7720-39-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.