Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminoimidazole | 7720-39-0 | MFCD00792530 | >98.0% | 83.09 g/mol | C3H5N3 | 1 ML
SDP

Catalog No. 5000210669
Encompass_Preferred

2-aminoimidazole is a small-molecule antibiofilm agent and a weak noncompetitive inhibitor of arginase I, supplied as a ready-to-use solution for research applications.

  • Provides 10 mM solution in DMSO, 1 mL volume.
  • CAS number 7720-39-0.
  • Purity >98.0%.
  • Molecular formula C3H5N3; molecular weight 83.09 g/mol.
  • Used to inhibit biofilm formation and modulate bacterial resistance mechanisms; arginase I inhibition Ki ≈ 3.6 mM.

Catalog No. 50-002-10669 Supplier Medchemexpress LLC Supplier No. HYW06221610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-aminoimidazole is a small-molecule antibiofilm agent and a weak noncompetitive inhibitor of arginase I, supplied as a ready-to-use solution for research applications.

  • Provides 10 mM solution in DMSO, 1 mL volume.
  • CAS number 7720-39-0.
  • Purity >98.0%.
  • Molecular formula C3H5N3; molecular weight 83.09 g/mol.
  • Used to inhibit biofilm formation and modulate bacterial resistance mechanisms; arginase I inhibition Ki ≈ 3.6 mM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.