Learn More
Medchemexpress LLC 2-[(aminoiminomethyl)amino]ethyl carbamimidothioic acid ester dihydrobromide | 100130-32-3 | 98.0% | 323.05 g/mol | C4H13Br2N5S | 5 MG
SDP
Catalog No. 5000241398
Encompass_Preferred
VUF 8430 dihydrobromide is a research-grade histamine H4 receptor agonist used in pharmacological and biochemical studies. It is supplied as a dihydrobromide salt with high reported potency and solubility, suitable for in vitro assays and receptor characterization.
  • Potent and selective H4 receptor agonist (Ki 31.6 nM; EC50 50 nM).
  • High purity of 98.0% for reproducible results.
  • Molecular weight 323.05 g/mol, white to off-white solid.
  • High water solubility (125 mg/mL) for assay preparation.
  • Available supporting documentation: data sheet, COA, SDS, NMR, MS.
Catalog No. 50-002-41398 Supplier Medchemexpress LLC Supplier No. HY1075555MG
VUF 8430 dihydrobromide is a research-grade histamine H4 receptor agonist used in pharmacological and biochemical studies. It is supplied as a dihydrobromide salt with high reported potency and solubility, suitable for in vitro assays and receptor characterization.
  • Potent and selective H4 receptor agonist (Ki 31.6 nM; EC50 50 nM).
  • High purity of 98.0% for reproducible results.
  • Molecular weight 323.05 g/mol, white to off-white solid.
  • High water solubility (125 mg/mL) for assay preparation.
  • Available supporting documentation: data sheet, COA, SDS, NMR, MS.