Learn More
Medchemexpress LLC 2-Aminoindan-2-phosphonic acid | 141120-17-4 | 213.17 | 10 MG
SDP
Catalog No. 5000445920
Encompass_Preferred
2-Aminoindan-2-phosphonic acid is an inhibitor of phenylalanine ammonia-lyase (PAL). It reduces the biosynthesis and accumulation of total phenolic compounds in the suspension cell culture system of *Cistanche*.
  • Inhibits phenylalanine ammonia-lyase (PAL) activity
  • Decreases accumulation of total phenolic compounds
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-45920 Supplier Medchemexpress LLC Supplier No. HYW00449410MG
2-Aminoindan-2-phosphonic acid is an inhibitor of phenylalanine ammonia-lyase (PAL). It reduces the biosynthesis and accumulation of total phenolic compounds in the suspension cell culture system of *Cistanche*.
  • Inhibits phenylalanine ammonia-lyase (PAL) activity
  • Decreases accumulation of total phenolic compounds
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month