Learn More
Medchemexpress LLC 2-Aminoindan-2-phosphonic acid | 141120-17-4 | 213.17 | 25 MG
SDP
Catalog No. 5000449384
Encompass_Preferred
2-Aminoindan-2-phosphonic acid acts as an inhibitor of phenylalanine ammonia-lyase (PAL), effectively reducing the biosynthesis and accumulation of total phenolic compounds within the suspension cell culture system of Cistanche. This compound is intended strictly for research purposes and is not for patient use.
  • Inhibits phenylalanine ammonia-lyase (PAL)
  • Reduces biosynthesis of phenolic compounds
  • Reduces accumulation of phenolic compounds
Catalog No. 50-004-49384 Supplier Medchemexpress LLC Supplier No. 25MGHYW004494
2-Aminoindan-2-phosphonic acid acts as an inhibitor of phenylalanine ammonia-lyase (PAL), effectively reducing the biosynthesis and accumulation of total phenolic compounds within the suspension cell culture system of Cistanche. This compound is intended strictly for research purposes and is not for patient use.
  • Inhibits phenylalanine ammonia-lyase (PAL)
  • Reduces biosynthesis of phenolic compounds
  • Reduces accumulation of phenolic compounds