Learn More
Medchemexpress LLC 2-aminoindan-2-phosphonic acid hydrochloride | 1416354-35-2 | MFCD23135861 | 98.4% | 249.63 g·mol⁻1 | C9H13ClNO3P | 100 MG
SDP
Catalog No. 5000299716
Encompass_Preferred
2-Aminoindan-2-phosphonic acid hydrochloride is a research-grade competitive phenylalanine ammonia lyase (PAL) inhibitor used in plant biology to decrease total phenolic compounds and phenylpropanoid glycoside (PheG) levels in cultured cells. It is supplied as a white to off-white solid with high reported purity and defined storage recommendations for long-term stability.
  • Competitive PAL inhibitor.
  • Reduces total phenolic compounds and PheG levels in plant cultured cells.
  • High purity suitable for biological research.
  • White to off-white solid form for ease of handling.
  • Defined storage stability in solvent and solid state supports reproducible results.
Catalog No. 50-002-99716 Supplier Medchemexpress LLC Supplier No. HYW017692100MG
2-Aminoindan-2-phosphonic acid hydrochloride is a research-grade competitive phenylalanine ammonia lyase (PAL) inhibitor used in plant biology to decrease total phenolic compounds and phenylpropanoid glycoside (PheG) levels in cultured cells. It is supplied as a white to off-white solid with high reported purity and defined storage recommendations for long-term stability.
  • Competitive PAL inhibitor.
  • Reduces total phenolic compounds and PheG levels in plant cultured cells.
  • High purity suitable for biological research.
  • White to off-white solid form for ease of handling.
  • Defined storage stability in solvent and solid state supports reproducible results.