Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminoindan-2-phosphonic acid hydrochloride | 1416354-35-2 | MFCD23135861 | 98.4% | 249.63 g·mol⁻¹ | C9H13ClNO3P | 250 MG
SDP

Catalog No. 5000303645
Encompass_Preferred

2-Aminoindan-2-phosphonic acid hydrochloride is a competitive phenylalanine ammonia lyase (PAL) inhibitor supplied as a hydrochloride salt for biochemical research; it is used to study phenolic compound biosynthesis and to modulate PAL-dependent pathways in plant and enzymatic assays.

  • High purity: 98.42%.
  • CAS number 1416354-35-2.
  • Chemical formula C9H13ClNO3P; molar mass 249.63 g·mol⁻¹.
  • Used to inhibit phenylalanine ammonia lyase (PAL) in plant and biochemical studies.
  • Available in small research quantities, such as 250 mg.

Catalog No. 50-003-03645 Supplier Medchemexpress LLC Supplier No. 250MGHYW017692
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Aminoindan-2-phosphonic acid hydrochloride is a competitive phenylalanine ammonia lyase (PAL) inhibitor supplied as a hydrochloride salt for biochemical research; it is used to study phenolic compound biosynthesis and to modulate PAL-dependent pathways in plant and enzymatic assays.

  • High purity: 98.42%.
  • CAS number 1416354-35-2.
  • Chemical formula C9H13ClNO3P; molar mass 249.63 g·mol⁻¹.
  • Used to inhibit phenylalanine ammonia lyase (PAL) in plant and biochemical studies.
  • Available in small research quantities, such as 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.