Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoindan hydrochloride, 98%

Catalog No. AC103860010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103860010 1 g Glass bottle
AC103860050 5 g Glass bottle
AC103860250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103860010 Supplier Thermo Scientific Chemicals Supplier No. 103860010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2338-18-3 | C9H11ClN | 168.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2338-18-3
Molecular Formula C9H11ClN
Molecular Weight (g/mol) 168.64
MDL Number MFCD00012549
InChI Key DLYRFJDJKGGVKT-UHFFFAOYSA-N
Synonym 2-aminoindan hydrochloride, 2-aminoindane hydrochloride, 2,3-dihydro-1h-inden-2-amine hydrochloride, 2-aminoindane hcl, 2-indanamine hydrochloride, 2-indanamine, hydrochloride, 2-indanylamine hydrochloride, su 8629 hydrochloride, indan-2-ylammonium chloride, indan-2-ylamine hydrochloride
PubChem CID 122764
IUPAC Name 2,3-dihydro-1H-inden-2-amine;hydrochloride
SMILES [Cl].NC1CC2=CC=CC=C2C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 246.0°C to 247.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 12,1196
Formula Weight 169.65
Percent Purity 98%
Chemical Name or Material 2-Aminoindan hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-048-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.