Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 2-(Aminomethyl)-1,3-dioxolane, >95%, [4388-97-0], J15276 103.12 C4H9NO2

Catalog No. 501409756
Encompass_Preferred

Compound: 2-(Aminomethyl)-1,3-dioxolane

CAS#: 4388-97-0

MDL#: not listed

Background: A dioxolane-protected aminoacetaldehyde used as a building block in pharmaceutical research. The primary amine allows for amide coupling, and the acetal can later be hydrolyzed to reveal the free aldehyde.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-9756 Supplier Frontier Specialty Chemicals Supplier No. J152761G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-(Aminomethyl)-1,3-dioxolane

CAS#: 4388-97-0

MDL#: not listed

Background: A dioxolane-protected aminoacetaldehyde used as a building block in pharmaceutical research. The primary amine allows for amide coupling, and the acetal can later be hydrolyzed to reveal the free aldehyde.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.