Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(aminomethyl)phenol

Catalog No. CDS008020 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008020 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008020 Supplier Sigma Aldrich Supplier No. CDS008020250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methyl-5-thiazoleethanol reacts with 4-ethyloctanoyl chloride to yield 4-methyl-5-thiazoleethanol 4-ethyloctanoate, sulfur-containing flavor compound
  • It was isolated from ether soluble alkaloidal fraction of panax ginseng.4-methyl-5-thiazoleethanol has been used in the synthesis of novel thiazolium halogenide ionic liquids.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 190675.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 932-30-9
Molecular Formula C7H9NO
Linear Formula C7H9NO
MDL Number MFCD00870498
Quantity 25 g
Molecular Weight (g/mol) 123.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.