Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Aminomethyl)pyrazine, 95%

Catalog No. AC454162500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454162500 250 mg
AC454160010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454162500 Supplier Thermo Scientific Chemicals Supplier No. 454162500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20010-99-5 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20010-99-5
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00673149
InChI Key HQIBSDCOMQYSPF-UHFFFAOYSA-N
Synonym 2-aminomethylpyrazine, 2-pyrazinemethanamine, 2-aminomethyl pyrazine, pyrazin-2-yl methanamine, 1-pyrazin-2-ylmethanamine, pyrazinemethanamine, 2-aminomethyl-pyrazine, pyrazin-2-ylmethylamine, pyrazin-2-ylmethyl amine
PubChem CID 266781
IUPAC Name pyrazin-2-ylmethanamine
SMILES NCC1=CN=CC=N1

Specifications

Boiling Point 87.0°C to 88.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 109.13
Percent Purity 95%
Chemical Name or Material 2-(Aminomethyl)pyrazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.