Learn More
Medchemexpress LLC (2-Aminophenyl)methanol (2-hydroxymethylaniline) | 5344-90-1 | 123.16 | 25 G
SDP
Catalog No. 5000449183
Encompass_Preferred
(2-Aminophenyl)methanol (2-Hydroxymethylaniline) is a molecular chaperone designed to rescue the P123S mutant pendrin. This compound offers advantages such as low dose, long-term effect, and low toxicity, making it suitable for research into Pendred syndrome, a type of syndromic deafness. It is intended for research use only.
  • Demonstrates significant rescue in HEK293 cells at 0.1 mM and maintains long-lasting efficacy after removal
  • Can be utilized for research into Pendred syndrome
  • Exhibits low dose, long-term effect, and low toxicity
  • Suitable for studying P123S mutant pendrin
Catalog No. 50-004-49183 Supplier Medchemexpress LLC Supplier No. 25GHYW004705
(2-Aminophenyl)methanol (2-Hydroxymethylaniline) is a molecular chaperone designed to rescue the P123S mutant pendrin. This compound offers advantages such as low dose, long-term effect, and low toxicity, making it suitable for research into Pendred syndrome, a type of syndromic deafness. It is intended for research use only.
  • Demonstrates significant rescue in HEK293 cells at 0.1 mM and maintains long-lasting efficacy after removal
  • Can be utilized for research into Pendred syndrome
  • Exhibits low dose, long-term effect, and low toxicity
  • Suitable for studying P123S mutant pendrin