Learn More
Medchemexpress LLC 2-Aminopurine | 452-06-2 | 99.6% | 135.13 | 100 MG
SDP
Catalog No. 5000453641
Encompass_Preferred
2-Aminopurine is a fluorescent analog of guanosine and adenosine, widely used as a fluorescence-decay-based probe of DNA structure. When 2-Aminopurine is inserted into an oligonucleotide, its fluorescence is highly quenched by stacking with natural bases. It has been utilized to probe nucleic acid structure and dynamics.
  • Fluorescent analog of guanosine and adenosine.
  • Widely used fluorescence-decay-based probe of DNA structure.
  • Its susceptibility to interbase quenching makes it an exquisitely sensitive fluorescent probe of nucleic acid structure.
  • Its fluorescence intensity is one thousand times that of adenine, despite differing only in the position of the exocyclic amine group.
Catalog No. 50-004-53641 Supplier Medchemexpress LLC Supplier No. HYW012642100MG
2-Aminopurine is a fluorescent analog of guanosine and adenosine, widely used as a fluorescence-decay-based probe of DNA structure. When 2-Aminopurine is inserted into an oligonucleotide, its fluorescence is highly quenched by stacking with natural bases. It has been utilized to probe nucleic acid structure and dynamics.
  • Fluorescent analog of guanosine and adenosine.
  • Widely used fluorescence-decay-based probe of DNA structure.
  • Its susceptibility to interbase quenching makes it an exquisitely sensitive fluorescent probe of nucleic acid structure.
  • Its fluorescence intensity is one thousand times that of adenine, despite differing only in the position of the exocyclic amine group.