Learn More
Medchemexpress LLC 2-Aminopurine-O-Ph-NHCO-C3-COOH hydrochloride | 95.3% | 420.85 | C18H21ClN6O4 | 5 MG
SDP
Catalog No. 5000281808
Encompass_Preferred
2-Aminopurine-O-Ph-NHCO-C3-COOH hydrochloride is a white to off-white solid used as the carboxylic acid precursor in the synthesis of activated esters. It is supplied with analytical purity and is intended for research and synthetic chemistry applications where a defined intermediate is required. Molecular formula C18H21ClN6O4; molecular weight 420.85 g/mol.
  • Used as a carboxylic acid intermediate for NHS-ester formation and other coupling reactions.
  • Supplied as a white to off-white solid suitable for synthetic workflows.
  • High reported purity of 95.3% for reliable reaction performance.
  • Available in small milligram quantities for research use.
  • Store sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-81808 Supplier Medchemexpress LLC Supplier No. HYW422359A5MG
2-Aminopurine-O-Ph-NHCO-C3-COOH hydrochloride is a white to off-white solid used as the carboxylic acid precursor in the synthesis of activated esters. It is supplied with analytical purity and is intended for research and synthetic chemistry applications where a defined intermediate is required. Molecular formula C18H21ClN6O4; molecular weight 420.85 g/mol.
  • Used as a carboxylic acid intermediate for NHS-ester formation and other coupling reactions.
  • Supplied as a white to off-white solid suitable for synthetic workflows.
  • High reported purity of 95.3% for reliable reaction performance.
  • Available in small milligram quantities for research use.
  • Store sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).