Learn More
Cayman Chemical 2-Aminopurine (hydrochloride), Purity: ≥98%, CAS: 1428126-74-2, 50 mg
SDP
Catalog No. 502965069
Encompass_Preferred
A fluorescent analog of guanosine and adenosine that is used as a site-specific probe of nucleic acid structure and dynamics, such as studies of base stacking interactions and DNA base flipping; ex/em = 320/381 nm, respectively, when adjusted to reduce interference with DNA base absorption as well as tryptophan fluorescence; inhibits PKR
Catalog No. 50-296-5069 Supplier Cayman Chemical Supplier No. 1860950 MG
A fluorescent analog of guanosine and adenosine that is used as a site-specific probe of nucleic acid structure and dynamics, such as studies of base stacking interactions and DNA base flipping; ex/em = 320/381 nm, respectively, when adjusted to reduce interference with DNA base absorption as well as tryptophan fluorescence; inhibits PKR