Learn More
Medchemexpress LLC 2-Aminoquinoline | 580-22-3 | 144.17 | 10 G
SDP
Catalog No. 5000447734
Encompass_Preferred
2-Aminoquinoline (2-Quinolinamine) is a compound that shows promise as a bioavailable neuronal nitric oxide synthase (nNOS) inhibitor. It also exhibits antiviral activity against the vaccinia virus, making it a potential subject for research into antineurodegenerative agents.
  • Functions as a bioavailable nNOS inhibitor
  • Demonstrates antiviral activity against vaccinia virus
  • Useful for research into antineurodegenerative agents
  • Has a purity of 99.95%
Catalog No. 50-004-47734 Supplier Medchemexpress LLC Supplier No. HYW00752410G
2-Aminoquinoline (2-Quinolinamine) is a compound that shows promise as a bioavailable neuronal nitric oxide synthase (nNOS) inhibitor. It also exhibits antiviral activity against the vaccinia virus, making it a potential subject for research into antineurodegenerative agents.
  • Functions as a bioavailable nNOS inhibitor
  • Demonstrates antiviral activity against vaccinia virus
  • Useful for research into antineurodegenerative agents
  • Has a purity of 99.95%