Learn More
Medchemexpress LLC 2-Aminoquinoline | 580-22-3 | 144.17 | 25 G
SDP
Catalog No. 5000449259
Encompass_Preferred
2-Aminoquinoline, also known as 2-Quinolinamine, is a promising compound primarily recognized as a bioavailable nNOS inhibitor. Despite exhibiting low human nNOS inhibition and selectivity against human eNOS, along with significant binding to other CNS targets, it demonstrates antiviral activity against the vaccinia virus. This compound holds potential for research in antineurodegenerative agents.
  • Acts as a bioavailable nNOS inhibitor
  • Shows antiviral activity against the vaccinia virus
  • Potential for research into antineurodegenerative agents
  • Appears as a solid
  • For research use only
Catalog No. 50-004-49259 Supplier Medchemexpress LLC Supplier No. 25GHYW007524
2-Aminoquinoline, also known as 2-Quinolinamine, is a promising compound primarily recognized as a bioavailable nNOS inhibitor. Despite exhibiting low human nNOS inhibition and selectivity against human eNOS, along with significant binding to other CNS targets, it demonstrates antiviral activity against the vaccinia virus. This compound holds potential for research in antineurodegenerative agents.
  • Acts as a bioavailable nNOS inhibitor
  • Shows antiviral activity against the vaccinia virus
  • Potential for research into antineurodegenerative agents
  • Appears as a solid
  • For research use only