Learn More
Medchemexpress LLC 2-Aminoquinoline | 580-22-3 | MFCD00006766 | 144.17 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0075241G
Encompass_Preferred
2-Aminoquinoline (2-Quinolinamine) is a promising compound being researched for its potential as an antineurodegenerative agent. It exhibits antiviral activity against the vaccinia virus. While it acts as a bioavailable nNOS inhibitor, it shows low human nNOS inhibition and limited selectivity against human eNOS, and it significantly binds to other CNS targets.
  • Acts as a bioavailable nNOS inhibitor
  • Exhibits antiviral activity against vaccinia virus
  • Potential for research into antineurodegenerative agents
Catalog No. 50-004-45942