Learn More
Medchemexpress LLC 2-Aminoterephthalic acid | 10312-55-7 | 181.15 | 1 ML
SDP
Catalog No. 5000448524
Encompass_Preferred
2-Aminoterephthalic acid is a biochemical reagent for life science research, functioning as a biological material or organic compound. It is utilized as a ligand for metal-organic frameworks (MOFs) in applications like gas adsorption and catalysis, and in copolymers for enhanced electrochemical activity. It also acts as a cross-linking agent to improve perovskite solar cell performance.
  • Acts as a ligand for metal-organic frameworks (MOFs) in gas adsorption, catalysis, and drug release.
  • Used in copolymers for enhanced electrochemical activity, cycling stability, and capacitance in capacitor manufacturing.
  • Enhances performance and stability of perovskite solar cells as a cross-linking agent.
Catalog No. 50-004-48524 Supplier Medchemexpress LLC Supplier No. HYW00467510MM/1ML
2-Aminoterephthalic acid is a biochemical reagent for life science research, functioning as a biological material or organic compound. It is utilized as a ligand for metal-organic frameworks (MOFs) in applications like gas adsorption and catalysis, and in copolymers for enhanced electrochemical activity. It also acts as a cross-linking agent to improve perovskite solar cell performance.
  • Acts as a ligand for metal-organic frameworks (MOFs) in gas adsorption, catalysis, and drug release.
  • Used in copolymers for enhanced electrochemical activity, cycling stability, and capacitance in capacitor manufacturing.
  • Enhances performance and stability of perovskite solar cells as a cross-linking agent.