Learn More
Medchemexpress LLC 2-aminoterephthalic acid | 10312-55-7 | 181.15 | 10 G
SDP
Catalog No. 5000448523
Encompass_Preferred
2-Aminoterephthalic acid is a biochemical reagent used in life science related research. It functions as an organic compound or biological material, serving as a versatile tool for various applications. It can be utilized in the formation of metal-organic frameworks (MOFs), which exhibit rich pore structures and high specific surface areas.
  • Ligand for the formation of metal-organic frameworks
  • Applicable in gas adsorption, catalysis, and drug release
  • Can be used for formation of copolymers with improved electrochemical activity, cycling stability, and capacitance
  • Suitable for capacitor manufacturing
  • Acts as a cross-linking agent to improve the performance and stability of perovskite solar cells
  • Passivates defects, enlarges grain size, and improves hydrophobicity
Catalog No. 50-004-48523 Supplier Medchemexpress LLC Supplier No. HYW00467510G
2-Aminoterephthalic acid is a biochemical reagent used in life science related research. It functions as an organic compound or biological material, serving as a versatile tool for various applications. It can be utilized in the formation of metal-organic frameworks (MOFs), which exhibit rich pore structures and high specific surface areas.
  • Ligand for the formation of metal-organic frameworks
  • Applicable in gas adsorption, catalysis, and drug release
  • Can be used for formation of copolymers with improved electrochemical activity, cycling stability, and capacitance
  • Suitable for capacitor manufacturing
  • Acts as a cross-linking agent to improve the performance and stability of perovskite solar cells
  • Passivates defects, enlarges grain size, and improves hydrophobicity