Learn More
Medchemexpress LLC 2-Aminoterephthalic acid | 10312-55-7 | MFCD00134536 | 181.15 | 25 G
SDP
Catalog No. 5000448007
Encompass_Preferred
2-Aminoterephthalic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It serves as a versatile compound in various applications including the formation of metal-organic frameworks, copolymers, and as a cross-linking agent.
  • Used as a ligand for the formation of metal-organic frameworks (MOFs) with metal ions, useful in gas adsorption, catalysis, and drug release.
  • Forms copolymers with improved electrochemical activity, cycling stability, and capacitance, suitable for capacitor manufacturing.
  • Acts as a cross-linking agent to improve the performance and stability of perovskite solar cells by passivating defects, enlarging grain size, and improving hydrophobicity.
Catalog No. 50-004-48007 Supplier Medchemexpress LLC Supplier No. 25GHYW004675
Add to Cart
Add to Cart
2-Aminoterephthalic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It serves as a versatile compound in various applications including the formation of metal-organic frameworks, copolymers, and as a cross-linking agent.
  • Used as a ligand for the formation of metal-organic frameworks (MOFs) with metal ions, useful in gas adsorption, catalysis, and drug release.
  • Forms copolymers with improved electrochemical activity, cycling stability, and capacitance, suitable for capacitor manufacturing.
  • Acts as a cross-linking agent to improve the performance and stability of perovskite solar cells by passivating defects, enlarging grain size, and improving hydrophobicity.