Learn More
Cayman Chemical 2-Aminotetralin, Purity: ≥95%, CAS: 2954-50-9, 500 mg
SDP
Catalog No. 502909872
Encompass_Preferred
A neuromodulatory agent; inhibits 5-HT and norepinephrine reuptake in rat brain at 39.4 mg/kg; induces hypothermia when administered by intracisternal or intraperitoneal injection; induces hyperthermia when implanted as crystals in the medial preoptic area of the hypothalamus in rats; substitutes for (+)-amphetamine in rats in a two-lever drug discrimination test in a dose-dependent manner
Catalog No. 50-290-9872 Supplier Cayman Chemical Supplier No. 35544500 MG
A neuromodulatory agent; inhibits 5-HT and norepinephrine reuptake in rat brain at 39.4 mg/kg; induces hypothermia when administered by intracisternal or intraperitoneal injection; induces hyperthermia when implanted as crystals in the medial preoptic area of the hypothalamus in rats; substitutes for (+)-amphetamine in rats in a two-lever drug discrimination test in a dose-dependent manner