Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminothiazole | 96-50-4 | MFCD00005325 | 100.14 g/mol | C3H4N2S | 10MM 1ML
SDP

Catalog No. 5000235284
Encompass_Preferred

Aminothiazole (2-aminothiazole) is a small heterocyclic amine used as a building block and research reagent in medicinal chemistry and chemical synthesis. It serves as a precursor for biologically active molecules including sulfur agents, biocides, fungicides, antibiotics, and dyes. The product is supplied in solution formats such as 10 mM in 1 mL and includes documented solubility and storage guidance for in vitro and in vivo use.

  • Cas number 96-50-4.
  • Molecular formula C3H4N2S; molecular weight 100.14 g/mol.
  • Soluble in water and dimethyl sulfoxide at high concentrations (e.g., ≥100 mg/mL in water, ≥50 mg/mL in DMSO).
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years); store solutions at -80°C (up to 2 years) or -20°C (up to 1 year).
  • Common applications include synthesis of pharmaceuticals, dyes, and agricultural agents.
  • Provided as a research reagent; follow appropriate handling and safety procedures.

Catalog No. 50-002-35284 Supplier Medchemexpress LLC Supplier No. HY1239610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aminothiazole (2-aminothiazole) is a small heterocyclic amine used as a building block and research reagent in medicinal chemistry and chemical synthesis. It serves as a precursor for biologically active molecules including sulfur agents, biocides, fungicides, antibiotics, and dyes. The product is supplied in solution formats such as 10 mM in 1 mL and includes documented solubility and storage guidance for in vitro and in vivo use.

  • Cas number 96-50-4.
  • Molecular formula C3H4N2S; molecular weight 100.14 g/mol.
  • Soluble in water and dimethyl sulfoxide at high concentrations (e.g., ≥100 mg/mL in water, ≥50 mg/mL in DMSO).
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years); store solutions at -80°C (up to 2 years) or -20°C (up to 1 year).
  • Common applications include synthesis of pharmaceuticals, dyes, and agricultural agents.
  • Provided as a research reagent; follow appropriate handling and safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.