Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminothiazole | 96-50-4 | MFCD00005325 | 99.9% | 100.14 g·mol⁻¹ | C3H4N2S | 500G
SDP

Catalog No. 5000235632
Encompass_Preferred

Aminothiazole (2-aminothiazole, CAS 96-50-4) is a heterocyclic amine used as a building block and reference compound in chemical and biochemical research. It serves as a precursor for the synthesis of biologically active molecules and is used in analytical and synthetic applications involving antibacterial, antifungal, and protease-targeted studies.

  • CAS number: 96-50-4.
  • Chemical formula: C3H4N2S.
  • Molecular weight: 100.14 g·mol⁻¹.
  • Purity: 99.9%.
  • Applications: synthetic intermediate and analytical reference compound.
  • Available pack sizes including 500 g.

Catalog No. 50-002-35632 Supplier Medchemexpress LLC Supplier No. HY12396500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Aminothiazole (2-aminothiazole, CAS 96-50-4) is a heterocyclic amine used as a building block and reference compound in chemical and biochemical research. It serves as a precursor for the synthesis of biologically active molecules and is used in analytical and synthetic applications involving antibacterial, antifungal, and protease-targeted studies.

  • CAS number: 96-50-4.
  • Chemical formula: C3H4N2S.
  • Molecular weight: 100.14 g·mol⁻¹.
  • Purity: 99.9%.
  • Applications: synthetic intermediate and analytical reference compound.
  • Available pack sizes including 500 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.