Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiazoline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A09365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09365G 5 g
A093625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09365G Supplier TCI America Supplier No. A09365G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3882-98-2
Molecular Formula C3H7ClN2S
Molecular Weight (g/mol) 138.613
MDL Number MFCD00012709
InChI Key CODJLHDDIDUKMD-UHFFFAOYSA-N
PubChem CID 520926
IUPAC Name 4,5-dihydro-1,3-thiazol-2-amine;hydrochloride
SMILES C1CSC(=N1)N.Cl

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 138.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazoline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.