Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amylpyridine 98.0+%, TCI America™
SDP

Catalog No. A227825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A227825G 25 g
A22785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A227825G Supplier TCI America Supplier No. A227825G

Chemical Identifiers

CAS 2294-76-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00051828
InChI Key HSDXVAOHEOSTFZ-UHFFFAOYSA-N
Synonym 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8
PubChem CID 16800
IUPAC Name 2-pentylpyridine
SMILES CCCCCC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 g
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Amylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.