Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Anilinopyridine, 97%, Thermo Scientific™

Catalog No. AAH57909MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57909MD 250 mg
AAH5790903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57909MD Supplier Thermo Scientific Chemicals Supplier No. H57909MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6631-37-4
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.22
MDL Number MFCD00006248
InChI Key HUDSSSKDWYXKGP-UHFFFAOYSA-N
Synonym 2-anilinopyridine, n-2-pyridylaniline, phenyl-2-pyridylamine, 2-phenylaminopyridine, n-phenyl-n-2-pyridinyl amine, 2-pyridineamine, n-phenyl, anilino-pyridine, phenylaminopyridine, 2-pyridylideneaniline, zlchem 1185
PubChem CID 81130
IUPAC Name N-phenylpyridin-2-amine
SMILES N(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 106°C to 112°C
Density 1.249 g/mL
Boiling Point 205°C to 206°C (7 mmHg)
Quantity 250 mg
Formula Weight 170.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Anilinopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-625-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.