Learn More
AdipoGen 2-Anthracenylsulfonyl chloride
SDP
Catalog No. 502561846
Encompass_Preferred
Chemical. CAS 17407-98-6. Formula C14H9ClO2S. MW 276.74. Synthetic. Derivatizing reagent with strong fluorescence.
Catalog No. 50-256-1846 Supplier AdipoGen Supplier No. CDXA0062M100
Chemical. CAS 17407-98-6. Formula C14H9ClO2S. MW 276.74. Synthetic. Derivatizing reagent with strong fluorescence.