Learn More
Enzo Life Sciences 2-Arachidonoylglycerol, (5mg), CAS Number: 53847-30-6
SDP
Supplier:  Enzo Life Sciences BMLFA0280005
Encompass_Preferred
Cannabinoid receptor ligand. Endogenous ligand for the CB1 receptor (CB1: Ki=58.3nM); CB2: Ki>3µM) present in high levels in the CNS. Alternative Name: 2-AG. Formula: C23H38O4. MW: 378.6. CAS Number: 53847-30-6. Purity: ≥98% (TLC). Appearance: Oil. Solubility: Soluble in DMSO or 100% ethanol. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -70°C. Stock solutions are stable for up to 3 months when stored at -70°C under an atmosphere of nitrogen or argon. Long Term Storage: -80°C
Catalog No. NC1762484