Learn More
Cayman Chemical 2-ArachIdonyl Glycerol-d5 1mg
SDP
Catalog No. 502912007
Encompass_Preferred
An internal standard for the quantification of 2-AG by GC- or LC-MS
Catalog No. 50-291-2007 Supplier Cayman Chemical Supplier No. 3621621 MG
An internal standard for the quantification of 2-AG by GC- or LC-MS