Learn More
Cayman Chemical Δ2-Avermectin B1a, Purity: ≥95%, 1 mg
SDP
Catalog No. 502933810
Encompass_Preferred
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 0.23 and 0.038 ppm, respectively)
Catalog No. 50-293-3810 Supplier Cayman Chemical Supplier No. 251191 MG
An ivermectin B1a degradation product; less active than ivermectin against T. urticae (LC90s = 0.23 and 0.038 ppm, respectively)